missing translation for 'onlineSavingsMsg'
Learn More

A 967079, Tocris Bioscience™

Product Code. 30001683
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective TRPA1 channel blocker

Selective TRPA1 channel blocker (IC50 values are 67 and 289 nM at human and rat TRPA1 receptors respectively). Displays 1000-fold selectivity for TRPA1 over other TRP channels, and is >150-fold selective over 75 other ion channels, enzymes and G-protein-coupled receptors. Exhibits analgesic effects in a rat osteoarthritic pain model.

Chemical Identifiers

CAS 1170613-55-4
Molecular Formula C12H14FNO
Molecular Weight (g/mol) 207.248
InChI Key HKROEBDHHKMNBZ-YAERWZOESA-N
Synonym 1e,3e-1-4-fluorophenyl-2-methyl-1-penten-3-one oxime
PubChem CID 122197682
IUPAC Name N-[(E)-1-(4-fluorophenyl)-2-methylpent-1-en-3-ylidene]hydroxylamine
SMILES CCC(=NO)C(=CC1=CC=C(C=C1)F)C

Specifications

Quantity 10 mg
Formula Weight 207.24
Percent Purity >98%
Chemical Name or Material A 967079
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'