Learn More
Tocris Bioscience™ A12B4C3
Potent and selective polynucleotide kinase/phosphatase (PNKP) inhibitor
Brand: Tocris Bioscience™ 5097/50
748.14 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
A12B4C3 is a potent and selective polynucleotide kinase/phosphatase (PNKP) inhibitor (IC50 = 60 nM). Displays no effect on calcineurin, protein PP-1 or APTX. Interrupts DNA repair and enhances radiosensitivity of human A549 lung carcinoma and MDA-MB-231 cells.Specifications
>98% | |
A12B4C3 | |
A12B4C3 |
50 mg | |
534.65 |
For Research Use Only