Learn More
Tocris Bioscience™ AA 147
ER proteostasis regulator
Brand: Tocris Bioscience™ 6759/5
67.29 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
AA 147 is an endoplasmic reticulum (ER) proteostasis regulator. Activates the ATF6 transcriptional program independently of ER stress and/or global UPR activation. Selectively reduces secretion of amyloidogenic transthyretin (TTR) from HepG2 cells. Reduces amyloidogenic immunoglobulin light chain (ALLC) secretion from ALMC-2 cells (EC50 = 1.1 μM).Specifications
N-(2-Hydroxy-5-methylphenyl)benzenepropanamide | |
5 mg | |
255.31 |
393121-74-9 | |
C16H17NO2 | |
98% |