missing translation for 'onlineSavingsMsg'
Learn More
Gibco™ AAV-MAX Control Plasmids Kit Product Code.: 17875057

Gibco™ AAV-MAX Control Plasmids Kit

Product Code. 17875057
1 Kit, 1 set
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17875057

Brand: Gibco™ A47672

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Gibco™ AAV-MAX Control Plasmids Kit consists of an optimized mix of three plasmids needed to produce adeno-associated virus serotype 2 (AAV2) expressing GFP: pAAV-CMV-emGFP, pAAV-Rep2Cap2, and pHelper.

The Gibco AAV-MAX Control Plasmids Kit consists of an optimized mix of three plasmids needed to produce adeno-associated virus serotype 2 (AAV2) expressing GFP: pAAV-CMV-emGFP, pAAV-Rep2Cap2, and pHelper. These three plasmids need to be used with a HEK293 cell line that stably expresses the adenovirus E1 gene. The kit contains an optimized mix of the three plasmids, as well as a separate tube of each of the three plasmids, which enables the user to utilize them in standalone manner and change the ratios.

• Saves process development time with optimized plasmid ratios
• Serves as a positive control for AAV2-GFP production with the AAV-MAX Helper-Free AAV Production system (Cat. No. A51217)

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
Product Type Plasmid
Product Line AAV-MAX
Quantity 1 Kit
Shipping Condition Ice
Format Kit
Regulatory Status Research Use Only (RUO)
Content And Storage • 1 tube containing mixture of all three plasmids, 1.5 mL
• 1 tube of each of the three plasmids alone, 0.75 mL

Store at –20°C.

Delivery Type Transfection
Expression System Mammalian

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number3096463 Certificate TypeCertificate of Analysis Date20/01/2025 Product CodeA47672
Lot Number3136525 Certificate TypeCertificate of Analysis Date14/01/2025 Product CodeA47672
Lot Number3096462 Certificate TypeCertificate of Analysis Date19/12/2024 Product CodeA47672
Lot Number3078329 Certificate TypeCertificate of Analysis Date15/10/2024 Product CodeA47672
Lot Number3070305 Certificate TypeCertificate of Analysis Date03/10/2024 Product CodeA47672

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ AAV-MAX Control Plasmids Kit >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.