missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ AAV-MAX Helper-Free AAV Production System Kit, without lysis buffer
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
- A scalable and high-titer platform for suspension AAV production
- Based on a high-density suspension culture of clonal 293F-derived Viral Production Cells 2.0 in Viral Production Medium
- Optimal viral titers are mediated by our advanced lipid nanoparticle transfection reagent in combination with a novel production enhancer
- The components are designed to work synergistically, resulting in maximal titers and eliminating the need to optimize reagents and protocols
- Note, this version of the kit does not contain the AAV-MAX Lysis Buffer
- To purchase the kit including the AAV-MAX Lysis Buffer, please see A51217
- All components are also available for purchase separately
Features of the AAV-MAX Helper-Free AAV Production System include:
- Components work synergistically to produce high AAV titers greater than 5x1010 vg/mL (unpurified)
- Suspension-based system has scalable protocols from shake flask to bioreactor scale
- Simplified workflow—streamlined protocol using helper virus-free triple transfection
- Clonal, 293F-derived, high-producer cell line
- Robust transfection protocols are adaptable to large scale transfection
Components of the AAV-MAX System Kit include:
- 1 vial of Viral Production Cells 2.0
- 1 L of Viral Production Medium
- 1 AAV-MAX Transfection Kit for 1 L of production
- 100 mL Viral-Plex Complexation Buffer
- 100 mL GlutaMAX™ Supplement
Specifications
Specifications
| Cell Line | Viral Production Cells 2.0 |
| Cell Type | Kidney (Embryonic) |
| Product Type | AAV Production System |
| Sterility | Sterile-filtered |
| For Use With (Application) | AAV production |
| Product Line | AAV-MAX |
| Quantity | 1 Kit |
| Content And Storage | • 1 vial Viral Production Cells 2.0 • 1 L Viral Production Medium • 1 AAV-MAX Transfection kit (for 1 L production) • 100 mL Viral-Plex Complexation Buffer • 100 mL GlutaMAX |
| Culture Type | Suspension Cell Culture |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ AAV-MAX Helper-Free AAV Production System Kit, without lysis buffer >
Spot an opportunity for improvement?Share a Content Correction