missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AAV-MAX pAAV-CMV-emGFP Vector

The Gibco™ AAV-MAX Control Plasmids can be used to produce adeno-associated virus serotype 2 (AAV2) expressing GFP.

Brand:  Gibco™ A47680

Product Code. 17895187

  • £608.00 / 0.75mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Gibco AAV-MAX Control Plasmids can be used to produce adeno-associated virus serotype 2 (AAV2) expressing GFP. The three plasmids include: pAAV-CMV-emGFP, a pAAV-Rep2Cap2, and pHelper. These three plasmids need to be used with a HEK293 cell line that stably expresses the adenovirus E1 gene. The pAAV-CMV-emGFP is a transfer plasmid, the pAAV-Rep2Cap2 is a Rep/Cap plasmid, and the pHelper is a helper plasmid.

• Standalone plasmids provide flexibility to optimize plasmid ratios
• When used together, serve as a positive control for AAV2-GFP production with the AAV-MAX Helper-Free AAV Production system (Cat. No. A51217)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 1 x 0.75 mL tube, 1 mg/mL

Store at -20°C.

1 mg/mL
Plasmid
AAV-MAX
Research Use Only (RUO)
Transfection
Mammalian
Non-sterile
1 Tube
Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ AAV-MAX pAAV-CMV-emGFP Vector > 0.75 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.