Learn More
Abamectin, Honeywell Fluka™
Brand: Honeywell Fluka R1781-100MG
Additional Details : CAS Number : 71751-41-2 Weight : 0.10000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : I
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Abamectin | |
≤ 100% | |
Avermectin B1 | |
White |
71751-41-2 | |
C48H72O14 (avermectin B1a) + C47H70O14 | |
155°C to 157°C | |
100 mg |