missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP1-59808
This item is not returnable.
View return policy
Description
ABCD2 Polyclonal specifically detects ABCD2 in Human samples. It is validated for Western Blot.Specifications
ABCD2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Adrenoleukodystrophy-like 1, Adrenoleukodystrophy-related protein, ALD1, ALDL1ATP-binding cassette sub-family D member 2, ALDRhALDR, ALDRPABC39, ATP-binding cassette, sub-family D (ALD), member 2 | |
Rabbit | |
Affinity purified | |
RUO | |
225 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9UBJ2 | |
ABCD2 | |
Synthetic peptides corresponding to ABCD2(ATP-binding cassette, sub-family D (ALD), member 2) The peptide sequence was selected from the middle region of ABCD2. Peptide sequence WRFEQLDTAIRLTLSEEKQKLESQLAGIPKMQQRLNELCKILGEDSVLKT. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Canine: 100%; Guinea pig: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Chicken: 85%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction