missing translation for 'onlineSavingsMsg'
Learn More

ABCD2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA5122297

Product Code. 17753694

  • £162.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ABCD2 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. The function of this peroxisomal membrane protein is unknown; however this protein is speculated to function as a dimerization partner of ABCD1 and/or other peroxisomal ABC transporters.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ABCD2
Polyclonal
Unconjugated
ABCD2
ABC39; Abcd2; adrenoleukodystrophy related; adrenoleukodystrophy-like 1; Adrenoleukodystrophy-related protein; ALD1; ALDL1; ALDR; ALDRP; ATP binding cassette subfamily D member 2; ATP-binding cassette sub-family D member 2; ATP-binding cassette, subfamily D (ALD), member 2; ATP-binding cassette, sub-family D (ALD), member 2; ATP-binding cassette, sub-family D, member 2; hALDR; LOW QUALITY PROTEIN: ATP-binding cassette sub-family D member 2
Rabbit
Antigen affinity chromatography
RUO
26874
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300
Q61285
ABCD2
Recombinant protein ATP Binding Cassette Transporter D2. The antigen corresponds to amino acid range 501-695 of the target protein.
20 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers