Learn More
Thermo Scientific™ Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates

384-well microarray plate with pyramidal well bottoms for maximum sample recovery.
£297.00 - £519.48
Specifications
Barcode | Custom Barcoding Available |
---|---|
Certifications/Compliance | Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant |
Format | Standard |
Description | 384-well Polypropylene Sample Processing & Storage Plates |
Array | 16 x 24 |
Product Code | Brand | Color | Volume (Metric) Well | Well Shape | Fit for Purpose | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Volume (Metric) Well | Well Shape | Fit for Purpose | Price | Quantity & Availability | ||||||
10701534
![]() |
Thermo Scientific™
AB-0781 |
Natural | 120 μL | Square, Conical Bottom (V-well) | PCR Ready |
£414.00
Pack of 50 |
In Stock
Log in to see stock. |
||||||
10324663
|
Thermo Scientific™
AB-0781/K |
Black | 120 μL | Square, Conical Bottom (V-well) | PCR Ready | N/A | N/A | N/A | |||||
10602674
![]() |
Thermo Scientific™
AB-1178 |
Natural | 250 μL | Square, Conical Bottom (V-well) | PCR Ready |
£356.00
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
11514764
![]() |
Thermo Scientific™
AB-0862 |
Natural | 300 μL | Square, Conical Bottom (V-well) | PCR Ready | N/A | N/A | N/A | |||||
10532255
![]() |
Thermo Scientific™
AB-1056 |
Natural | 55 μL | Round, Conical Bottom (V-well) | PCR Ready |
£297.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Thermo Scientific™ Abgene™ polypropylene microplates and DeepWell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use and have molded slots and pads for automated sample processing and preparation workflows. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using USP <661.1> high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and DeepWell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
All Thermo Scientific plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.
Highlights
- Certified free from DNAse, RNAse, PCR Inhibitors, and human DNA
- Pyramidal well bottom for ease of sample retrieval
- Discrete raised rims around each well for effective sealing and to reduce the risk of cross-contamination
Compatible Sealing Film: Adhesive and Heat
Hazardous: No
Max Centrifugal Force: 2000-5000 RCF
Storage Condition: Ambient stored, or transported
All plates are ANSI format for compatibility with automated systems.
To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.
Specifications
Custom Barcoding Available | |
Standard | |
16 x 24 | |
Low binding PP | |
384-well plate | |
Polypropylene | |
Cleanroom | |
Yes | |
5000 x g | |
For Research Use Only. Not for use in diagnostic procedures. | |
PCR Ready |
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
384-well Polypropylene Sample Processing & Storage Plates | |
Autoclavable | |
Abgene | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
384 | |
Non-treated | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
50 | |
Yes | |
ANSI/ SLAS Standard |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.