Learn More
Abietic acid, 90+%, Thermo Scientific Chemicals
£88.30 - £309.00
Chemical Identifiers
CAS | 514-10-3 |
---|---|
Molecular Formula | C20H30O2 |
Molecular Weight (g/mol) | 302.46 |
MDL Number | MFCD03423567 |
InChI Key | RSWGJHLUYNHPMX-ONCXSQPRSA-N |
Synonym | abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183 |
PubChem CID | 10569 |
ChEBI | CHEBI:28987 |
IUPAC Name | (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid |
SMILES | CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11369198
|
Thermo Scientific Alfa Aesar
042582.MD |
250 mg |
£88.30
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11379198
|
Thermo Scientific Alfa Aesar
042582.03 |
1 g |
£309.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Abietic acid is used to make ester gums in lacquers, varnishes and soaps. It is also used in metal resonates and plastics. Further, it is utilized in musical instruments to make them less slippery. It acts as a precursor to natural product synthesis. In addition to this, it is used to depackage integrated circuits from their epoxy coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
514-10-3 | |
302.46 | |
RSWGJHLUYNHPMX-ONCXSQPRSA-N | |
10569 | |
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid |
C20H30O2 | |
MFCD03423567 | |
abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183 | |
CHEBI:28987 | |
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1 |
Specifications
514-10-3 | |
Yellow | |
MFCD03423567 | |
14,7 | |
Soluble in acetone,petroleum ether,diethyl ether and ethanol. Insoluble in water. | |
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1 | |
302.46 | |
CHEBI:28987 | |
≥90% | |
Crystalline | |
Abietic acid |
148°C to 152°C | |
C20H30O2 | |
2221451 | |
abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183 | |
RSWGJHLUYNHPMX-ONCXSQPRSA-N | |
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid | |
10569 | |
302.45 | |
Characteristic | |
Air sensitive |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 208-178-3
missing translation for 'rtecsNumber' : TP8580000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : 2°C to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
- Abietic acid
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Abietic acid
RUO – Research Use Only