missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abietic acid, tech. 75%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Abietic acid is a useful precursor for synthesis of various natural products including diterpenes and steroids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Abietic acid is a useful precursor for synthesis of various natural products including diterpenes and steroids.
Solubility
Soluble in alcohols, acetone and ethers
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Store under inert gas. Sensitive to air.
Abietic acid is a useful precursor for synthesis of various natural products including diterpenes and steroids.
Solubility
Soluble in alcohols, acetone and ethers
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Store under inert gas. Sensitive to air.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Beilstein | 2221451 |
Merck Index | 14,7 |
Solubility Information | Soluble in alcohols,acetone and ethers |
Formula Weight | 302.45 |
Percent Purity | 75% |
Grade | Technical |
Quantity | 25 g |
Odor | Characteristic |
Sensitivity | Air sensitive |
Chemical Name or Material | Abietic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'