Learn More
Abietic acid, tech. 75%, Thermo Scientific Chemicals
£29.75 - £82.51
Chemical Identifiers
CAS | 514-10-3 |
---|---|
Molecular Formula | C20H30O2 |
Molecular Weight (g/mol) | 302.46 |
MDL Number | MFCD03423567 |
InChI Key | RSWGJHLUYNHPMX-ONCXSQPRSA-N |
Synonym | abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183 |
PubChem CID | 10569 |
ChEBI | CHEBI:28987 |
IUPAC Name | (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid |
SMILES | CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11303237
|
Thermo Scientific Alfa Aesar
L13770.14 |
25 g |
£29.75
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11313237
|
Thermo Scientific Alfa Aesar
L13770.22 |
100 g |
£82.51
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Abietic acid is a useful precursor for synthesis of various natural products including diterpenes and steroids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAbietic acid is a useful precursor for synthesis of various natural products including diterpenes and steroids.
Solubility
Soluble in alcohols, acetone and ethers
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Store under inert gas. Sensitive to air.
Chemical Identifiers
514-10-3 | |
302.46 | |
RSWGJHLUYNHPMX-ONCXSQPRSA-N | |
10569 | |
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid |
C20H30O2 | |
MFCD03423567 | |
abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183 | |
CHEBI:28987 | |
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1 |
Specifications
514-10-3 | |
MFCD03423567 | |
14,7 | |
Soluble in alcohols,acetone and ethers | |
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1 | |
302.46 | |
CHEBI:28987 | |
75% | |
Characteristic | |
Abietic acid |
C20H30O2 | |
2221451 | |
abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183 | |
RSWGJHLUYNHPMX-ONCXSQPRSA-N | |
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid | |
10569 | |
302.45 | |
Technical | |
Air sensitive |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 208-178-3
missing translation for 'rtecsNumber' : TP8580000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only