missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ABL2 HEK293 Cell Transient Overexpression Lysate (Nondenatured)

Human overexpression lysate

Brand:  Abnova™ L022T6.100ug


Product Code. 16170884

  • £182.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

HEK293
ABL2 HEK293 Cell Transient Overexpression Lysate(Non-Denatured)
Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
pCMV-ABL2 full length
100 μg
Human
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Immunoprecipitation, Western Blot
BC065912
Theoretical M.W. 127kDa
Transient overexpression cell lysate was tested with anti-ABL2 antibody (Mfr. No. H00000027-M05) by Western Blot.
In modified RIPA Lysis Buffer.
2 mg/mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 100UG ABL2 HEK293 Cell Transient Overexpression Ly

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Mercaptoethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers