missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Blood/Cultured Cell Total RNA Purification Kit

Blood/Cultured Cell Total RNA Purification Kit is designed specifically for purifying total RNA from fresh whole human blood and cultured animal cells.

Brand:  Abnova™ U0064.1Kit

Product Code. 16032378

  • £577.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Kit was designed specifically for purifying total RNA from fresh whole human blood and cultured cells
  • Entire procedure can be completed within 30 minutes
  • Applications include RT-PCR, Northern Blotting, Primer Extension, mRNA Selection, and cDNA Synthesis
  • Yield of 5-30 μg

Specifications

Specifications

Store at room temperature.
Purification
Blood/Cultured Cell Total RNA Purification Kit
Blood/Cultured Cell Total RNA Purification Kit is designed specifically for purifying total RNA from fresh whole human blood and cultured animal cells.
200mL RBC Lysis Buffer, 60mL RB Buffer, 30mL RT Buffer, 50mL W1 Buffer, 25mL Wash Buffer (add 100mL Ethanol), 6mL RNase-free Water, RB column, 2mL collection tube
1 Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers