missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Cultured Cell Total RNA Purification Kit

Designed specifically for purifying total RNA from cultured animal cells

Brand:  Abnova™ U0067.1Kit

Product Code. 16062378

  • £577.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Sample Type: Cultured cell
  • Sample Size: Cultured animal cells (up to 5 x 106)

Purification

Specifications

Specifications

Store at room temperature.
Purification
Cultured Cell Total RNA Purification Kit
Cultured Cell Total RNA Purification Kit was designed specifically for purifying total RNA from cultured animal cells.
60mL RB Buffer, 130mL W1 Buffer, 50mL Wash Buffer (add 200mL Ethanol), 30mL RNase-free Water, RNA binding plate, 0.35mL collection plate, adhesive film
1 Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers