Learn More
Abnova™ Fluorescent Dye 555-C3 Azide
Cyanine 3 dyes have enhanced fluorescence upon binding to proteins; C3 Azide reacts with alkyne groups
Brand: Abnova™ U0200.1mg
Additional Details : Weight : 0.00010kg
Description
Ideal for labeling of biological molecules (e.g. peptides, proteins, oligoes) for fluorescence imaging and other fluorescence-based biochemical analysis
- Chemical Formula: C35H43F3N6O9S2
- Mol. Weight: 812.88g/mol
- Purity: ≥90%
- Molar Extinction Co-efficient: ≥1,00,000 M-1cm-1
- Excitation (Max): 555nm
- Emission (Max): 565nm
- Solubility: DMSO, water
Application: Conjugation, In vivo imaging
Recommended Storage: Store in the dark at -20°C.
Specifications
Cyanine 3 dyes have enhanced fluorescence upon binding to proteins. C3 Azide reacts with alkyne groups. | |
1 mg | |
812.88g/mol | |
Store in the dark at -20°C | |
565 |
Conjugation; in vivo Imaging | |
Fluorescent Dye 555-C3 Azide | |
≥90% | |
Fluorescent Dye 555-C3 Azide | |
555 |
For research use only (RUO).