Learn More
Abnova™ Fluorescent Dye 678-C5.5 bisAcid
Cyanine 5.5 dyes are common red fluorophores
Brand: Abnova™ U0225.5mg
Additional Details : Weight : 0.00010kg
Description
Ideal for labeling of biological molecules (e.g. peptides, proteins, oligoes) for fluorescence imaging and other fluorescence-based biochemical analysis
- Chemical Formula: C47H51F3N2O18S4
- Mol. Weight: 1117.17g/mol
- Purity: ≥90%
- Molar Extinction Co-efficient: ≥1,00,000 M-1cm-1
- Excitation (Max): 678nm
- Emission (Max): 701nm
- Solubility: DMSO, water
Application: Conjugation, In vivo imaging
Recommended Storage: Store in the dark at -20°C.
Specifications
Cyanine 5.5 dyes are common red fluorophores. | |
5 mg | |
1117.17g/mol | |
Store in the dark at -20°C | |
701 |
Conjugation; in vivo Imaging | |
Fluorescent Dye 678-C5.5 bisAcid | |
≥90% | |
Fluorescent Dye 678-C5.5 bisAcid | |
678 |
For research use only (RUO).