Learn More
Abnova™ Fluorescent Dye 749-C7 Azide
Cyanine 7 dyes are common near infrared red fluorophores used in in vivo imaging applications; C7 Azide reacts with alkyne groups
Brand: Abnova™ U0219.1mg
Additional Details : Weight : 0.00010kg
Description
Ideal for labeling of biological molecules (e.g. peptides, proteins, oligoes) for fluorescence imaging and other fluorescence-based biochemical analysis
- Chemical Formula: C39H47F3N6O9S2
- Mol. Weight: 997.11g/mol
- Purity: ≥90%
- Molar Extinction Co-efficient: ≥1,00,000 M-1cm-1
- Excitation (Max): 749nm
- Emission (Max): 776nm
- Solubility: DMSO, water
Application: Conjugation, In vivo imaging
Recommended Storage: Store in the dark at -20°C.
Specifications
Cyanine 7 dyes are common near infrared red fluorophores used in in vivo imaging applications. C7 Azide reacts with alkyne groups. | |
1 mg | |
997.11g/mol | |
Store in the dark at -20°C | |
776 |
Conjugation; in vivo Imaging | |
Fluorescent Dye 749-C7 Azide | |
≥90% | |
Fluorescent Dye 749-C7 Azide | |
749 |
For research use only (RUO).