Learn More
Abnova™ Fluorescent Dye 750-I Hydrazide
Water-soluble fluorescent dye forms stable Schiff Base with carbonyl group
Brand: Abnova™ U0261.1mg
Additional Details : Weight : 0.00010kg
Description
Compared with Alexa Fluor 750, Fluorescent Dye 750-I is much brighter with absorption at 633nm. Fluorescence emission of Fluorescent Dye 750-I Hydrazide is well separated from that of other commonly used red fluorophores, such as TAMRA, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 750, Cy7, DyLight 755
- Purity: >90%
- Molar Extinction Co-efficient: >2,40,000 M-1cm-1
- Excitation (Max): 749nm
- Emission (Max): 775nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Fluorescent dye that is water-soluble and forms stable Schiff Base with carbonyl group. Compared with Alexa Fluor 750, Fluorescent Dye 750-I is much brighter with absorption at 633nm. Fluorescence emission of Fluorescent Dye 750-I Hydrazide is well separated from that of other commonly used red fluorophores, such as TAMRA, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. | |
1 mg | |
>90% | |
DMSO | |
Fluorescent Dye 750-I Hydrazide | |
749 |
Conjugation; Flow Cytometry | |
Fluorescent Dye 750-I Hydrazide | |
Alexa Fluor 750, Cy7, DyLight 755 | |
Store the product in desiccated environment at -20°C and avoid from light. | |
775 |
For research use only (RUO).