missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Fluorescent Dye Eu-Streptavidin

Fluorescent Dye Eu with Streptavidin conjugate

Brand:  Abnova™ U0310.100ug

Additional Details : Weight : 0.00010kg

Product Code. 16294960

  • £464.00 / 100µg
Estimated Shipment: 14-08-2024
to see stock.
Add to basket

Description

Description

Fluorescent Dye Eu is optimized for TR-FRET applications with large Stokes shifts and extremely long emission half-lives when compared to traditional fluorophores such as Alexa Fluor™ or Cyanine dyes. Ideal for labeling via Streptavidin for fluorescence imaging and other fluorescence-based biochemical analysis. For most fluorescent streptavidin applications, a concentration of 1 to 20g/mL is sufficient. The optimal working dilution should be determined by the end user.

  • Typical Acceptor for TR-FRET: Alexa 647, APC, Cy5, DyLight 650, Fluorescence Dye 647-I, TF5
  • Molar Extinction Co-efficient: ≥22,000 M-1cm-1
  • Excitation (Max): 346nm
  • Emission (Max): 617nm
  • Solubility: Water
  • Storage Buffer: Water

Application: Conjugation, Time Resolved-Fluorescence Resonance Energy Transfer

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Specifications

Fluorescent Dye Eu with Streptavidin conjugate. Fluorescent Dye Eu is optimized for TR-FRET applications with large Stokes shifts and extremely long emission half-lives when compared to traditional fluorophores such as Alexa Fluor™ or Cyanine dyes.
100 μg
Alexa 647, APC, Cy5, DyLight 650, Fluorescence Dye 647-I, TF5
Store the product in desiccated environment at -20°C and avoid from light. Add ddH2O with 0.1% BSA into the vial to have 1 mg/mL stock solution. This reconstituted solution remains stable for 2 months when stored in the presence of 2 mM sodium azide and avoid from light at 4°C. For longer storage, divid the reconstituted solution into single-used aliquots, or add equal volume of glycerol without aliquoting, and avoid from light at -20°C.
617
Conjugation; Time Resolved-Fluorescence Resonance Energy Transfer
Fluorescent Dye Eu-Streptavidin
Water
Fluorescent Dye Eu-Streptavidin
346
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only (RUO).