missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Plant Total RNA Purification Kit
Plant Total RNA Purification Kit is designed specifically for purifying total RNA from plant tissue and cells.
Brand: Abnova™ U0065.1Kit
Additional Details : Weight : 0.02000kg
Description
- Provides an efficient method for purifying total RNA from plant tissue and cells
- Samples are ground in liquid nitrogen and then filtered to remove cell debris
- In the presence of a binding buffer and chaotropic salt, total RNA in the lysate binds to the glass fiber matrix of the spin column
- Optional DNase treatments can be followed to remove unwanted DNA residue
- The procedure does not require phenol extraction or alcohol precipitation and can be completed within 30 minutes
- The purified total RNA is ready for use in RT, RT-PCR, Real-time PCR and Northern Blotting
- Store at room temperature
Specifications
Store at room temperature. | |
Purification | |
Plant Total RNA Purification Kit |
Plant Total RNA Purification Kit is designed specifically for purifying total RNA from plant tissue and cells. | |
60mL RB Buffer, 60mL PRB Buffer, 50mL W1 Buffer, 25mL Wash Buffer (add 100mL Ethanol), 15mL RNase-free Water, filter column, RB column, 2mL collection tube | |
1 Kit |