missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Tissue Total RNA Purification Kit
Tissue Total RNA Purification Kit is designed specifically for purifying total RNA from a variety of animal and paraffin-embedded tissue.
Brand: Abnova™ U0066.1Kit
Additional Details : Weight : 0.02000kg
Description
- Tissue samples can be efficiently homogenized in a microcentrifuge tube using the provided micropestle
- Detergents and chaotropic salt are used to lyse cells and inactivate RNase with an optional in-column DNase treatment
- The procedure does not require phenol extraction or alcohol precipitation and can be completed within 15 minutes
- The purified RNA is ready for use in RT-PCR, Northern Blotting, Primer Extension and cDNA Library Construction
- Stores dry at room temperature (15-25°C) for at least 9 months without showing any reduction in performance
- For research use only
Specifications
Store at room temperature. | |
Purification | |
Tissue Total RNA Purification Kit |
Tissue Total RNA Purification Kit is designed specifically for purifying total RNA from a variety of animal and paraffin-embedded tissue. | |
60mL RB Buffer, 50mL W1 Buffer, 25mL Wash Buffer (add 100mL Ethanol), 15mL RNase-free Water, filter column, RB column, 2mL collection tube, micropestle | |
1 Kit |