Learn More
Novus Biologicals™ AbPure™ Antibody Concentration and Clean-Up Antibody Purification Kit
Allows removal of contaminants that interfere in conjugation reactions of InnovaCoat Gold
Brand: Novus Biologicals™ 262-0010
Additional Details : Weight : 0.58200kg
Description
Kit Components- 3 spin cartridge/collecting tube assemblies
- 1 bottle of AbPure Conjugation Buffer
Specifications
4°C | |
3 spin cartridge/collecting tube assemblies, 1 bottle of AbPure Conjugation Buffer | |
AbPure Antibody Concentration and Clean-Up Antibody Purification Kit |
3 Tests | |
3 columns |
For research use only.