Learn More
Novus Biologicals™ ABpure™ Antibody Purification Kit
Novus Biologicals™ ABpure Antibody Purification Kit is used to remove contaminants such as BSA and other proteins, glycine, tris, and azide that interfere in conjugation reactions.
Brand: Novus Biologicals™ 260-0005
Additional Details : Weight : 0.58200kg
Description
- Used to purify antibodies from crude samples such as ascites fluid or immune serum
- Antibody to be purified or cleaned up ideally is in volume of 0.1mL to 0.5mL
- 20 to 500μg of antibody can be purified in each run
Specifications
4°C | |
4 or 12 additional collecting tubes, 1 vial AbPure wash buffer, 1 vial AbPure eution buffer, 1 vial AbPure neutralization buffer, 1 or 3 vials Protein A resin, 1 vial 10X AbPure binding buffer, 1/3 spin cartridge/collecting tube assemblies | |
Antibody Purification Kit |
1 Test | |
One purification |
For research use only.