missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ABpure™ Antibody Purification Kit

Novus Biologicals™ ABpure Antibody Purification Kit is used to remove contaminants such as BSA and other proteins, glycine, tris, and azide that interfere in conjugation reactions.

Brand:  Novus Biologicals™ 260-0010

Additional Details : Weight : 0.58200kg

 View more versions of this product

Product Code. 18099633

  • £703.84 / 3 tests
Estimated Shipment: 19-08-2024
to see stock.

Description

Description

  • Used to purify antibodies from crude samples such as ascites fluid or immune serum
  • Antibody to be purified or cleaned up ideally is in volume of 0.1mL to 0.5mL
  • 20 to 500μg of antibody can be purified in each run
Specifications

Specifications

4°C
1 or 3 spin cartridge/collecting tube assemblies, 1 vial 10X AbPure binding buffer, 1 vial AbPure neutralization buffer, 1 vial AbPure wash buffer, 1 vial AbPure elution buffer, 4 or 12 additional collecting tubes, 2 or 3 vials of Protein A resin
Antibody Purification Kit
3 Tests
Three purifications
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only.