Learn More
Novus Biologicals™ AbPure™ BSA Removal Antibody Purification Kit
Novus Biologicals™ AbPure BSA Removal Kit can be used on any antibody sub-type, and species.
Brand: Novus Biologicals™ 263-0100
Additional Details : Weight : 0.58200kg
Description
Kit Components:- 1 x vial AbPure BSA Removal Buffer
- 1 x vial AbPure Re-Suspension Buffer
Specifications
4°C | |
1 mL | |
1 x vial AbPure BSA Removal Buffer, 1 x vial AbPure Re-Suspension Buffer | |
AbPure BSA Removal Kit |
For research use only.