missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AbPure™ Magnetic Beads Antibody Purification Kit
Novus Biologicals™ AbPure Magnetic Antibody Purification Kit is designed to be compatible with Lightning-Link, Thunder-Link PLUS, InnovaCoat GOLD and LATEX conjugation kits.
Brand: Novus Biologicals™ 265-0200
Additional Details : Weight : 0.58200kg
Description
Kit Components:- 400μL of Protein A Magnetic Beads
- One vial of Neutralization Buffer
- One bottle of Wash Buffer
- One vial of 10X Binding Buffer
- One vial of Elution Buffer
- Not supplied magnetic stand
Specifications
4°C | |
400μL Protein A Magnetic Beads, 1 vial of neutralization buffer, 1 bottle of wash buffer, 1 vial of 10X binding buffer, 1 vial of elution buffer, Not supplied magnetic stand | |
AbPure Magnetic Antibody Purification Kit |
0.4 mL | |
One reaction |
For research use only.