missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ABpure™ Mouse Antibody Purification Kit
Novus Biologicals™ ABpure Mouse Antibody Purification Kit is used to remove contaminants such as BSA and other proteins, glycine, tris, and azide that interfere in conjugation reactions.
£385.12 - £687.24
Specifications
Content And Storage | 4°C |
---|---|
Product Type | Antibody Purification Kit |
Alert:
Not suitable for use with antibodies from other species
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
18061004
|
Novus Biologicals™
261-0010 |
Three purifications |
£687.24
3 tests |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18093544
|
Novus Biologicals™
261-0005 |
One purification |
£385.12
1 test |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Used to purify antibodies from crude samples such as ascites fluid or immune serum
- Antibody to be purified or cleaned up ideally is in volume of 0.1mL to 0.5mL
- 20 to 150μg of antibody can be purified in each run
Specifications
4°C | |
Antibody Purification Kit |
For research use only.