missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Slide Dispenser

Obtain finger print free microscope slides with ease. Kartell™ Slide Dispenser uses a rotating knob to release one of up to 50 dust protected slides.

Brand:  Kartell™ 0054000

141.56 GBP valid until 2024-11-29
Use promo code "21706" to get your promotional price.


Product Code. 10325681

  • £194.00 / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For finger print free dispensing of microscope slides
  • Up to 50 slides can be put in the magazine
  • A cover protects the slides from dust
  • Each slide is dispensed by rotating the knurled knob on either side of the moulded base
  • Rubber rollers at the sides of the slides ease out the slide which can then be handled by the edges
Specifications

Specifications

50
100 x 120 x 140 mm
ABS, Polystyrene
Slides
140 mm
76 x 26 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers