Learn More
Novus Biologicals™ AbSelect™ G Serum Antibody Purification Kit
For purification of antibodies from crude samples such as ascites fluid or immune serum
£575.47 - £1216.23
Specifications
Product Type | Antibody Purification Kit |
---|---|
Content And Storage | 4°C. Do not freeze. |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
18065704
|
Novus Biologicals™
893-0030 |
1 Purification |
£575.47
1 test |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18076004
|
Novus Biologicals™
893-0500 |
3 Purifications |
£1216.23
3 tests |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Protein G has a high affinity for the Fc regions of IgG molecules from a variety of species. AbSelect G Serum resin is prepared by coupling purified Protein G to agarose beads. It can therefore be used to purify IgG fractions from both serum and ascites fluid.
- Compatible with the lightning-link antibody labeling kits
- Allows purified antibody to be immediately labeled with an enzyme, fluorescent protein or fluorescent dye, with a hands-on time of under 30 seconds
Specifications
Antibody Purification Kit | |
4°C. Do not freeze. |