missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AbSelect™ Mouse Antibody Purification Kit
For purification of mouse antibodies from crude samples such as ascites fluid
£440.45 - £710.48
Specifications
Product Type | Antibody Purification Kit |
---|---|
Content And Storage | 4°C. Do not freeze. |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
18023224
|
Novus Biologicals™
830-0005 |
1 Purification |
£440.45
1 test |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18073494
|
Novus Biologicals™
830-0010 |
3 Purifications |
£710.48
3 tests |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Mouse antibody to be purified or cleaned up is in volume of 0.1 to 0.5mL
- Up to 150g of antibody can be purified in each run
- This system is not suitable for use with antibodies from other species
- Components of the AbSelect kit are fully compatible with lightning-link and lightning-link rapid antibody labeling system
Specifications
Antibody Purification Kit | |
4°C. Do not freeze. |