missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AbSelect™ Mouse TCS Antibody Purification Kit
For purification of mouse IgG fractions from hybridoma supernatants
£458.95 - £901.70
Specifications
Product Type | Antibody Purification Kit |
---|---|
Content And Storage | 4°C. Do not freeze. |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
18721144
|
Novus Biologicals™
832-0005 |
1 Purification |
£458.95
1 test |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18747713
|
Novus Biologicals™
832-0500 |
3 Purifications |
£901.70
3 tests |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
AbSelect Mouse TCS resin has a high affinity for mouse IgG molecules. The method involves capture of the antibody on the AbSelect Mouse TCS resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.
- This system is not suitable for use with antibodies from other species
- Binding strength of Bovine IgG to AbSelect Mouse resin is negligible
- Components of the AbSelect kit are fully compatible with lightning-link and lightning-link rapid antibody labeling system
Specifications
Antibody Purification Kit | |
4°C. Do not freeze. |