Learn More
Novus Biologicals™ AbSelect™ Serum Antibody Purification Kit
Fast and simple method to purify antibodies from ascites fluid and serum
Brand:  Novus Biologicals™ 863-0500
Additional Details : Weight : 0.58200kg
 View more versions of this product
Product Code. 18797083
  • £836.91 / 3 tests
Estimated Shipment: 19-08-2024
to see stock.
Description
Description
AbSelect Serum is prepared by coupling highly purified protein A to agarose beads which can be used to purify IgG fractions from ascites fluid and serum. The method involves the capture of the antibody on the AbSelect Serum resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.
  •  Compatible with the lightning-link antibody labeling kits
  • Allows purified antibody to be immediately labeled with an enzyme, fluorescent protein or fluorescent dye
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
Specifications
Antibody Purification Kit
3 Purifications
4°C. Do not freeze.
3 Tests