missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABsolute Blue QPCR Mix, ROX

ABsolute Blue qPCR Mixes utilize Thermo-Start Taq hot start enzyme for increased sensitivity.

Brand:  Thermo Scientific™ AB4139A

223.50 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11872153

  • £447.00 / 400 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ ABsolute Blue qPCR Mixes contain an inert blue dye to significantly enhance the contrast between reagent and plastic, allowing quick and easy verification that master mix has been dispensed.

  • High performance - ABsolute sensitivity, consistency & convenience
  • Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
  • Universal - compatible with all major qPCR platforms
  • Adaptable - available with different ROX passive reference dye concentrations
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
• 1 x 5 mL ABsolute Blue qPCR ROX Mix (2X)
Sufficient for 400 x 25 μL reactions.

Store at -20°C
Tube
qPCR
Standard
ABsolute Blue
400 Reactions
2 X
DNA, dsDNA
Primer-probe
High
Taq DNA Polymerase
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
qPCR Mix
2X
400 Reactions
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.