missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABsolute QPCR Mix, ROX

Optimized for use with hybridization probes and contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes.

Brand:  Thermo Scientific™ AB1139A

 View more versions of this product

Product Code. 11857682

  • £456.00 / 400 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Enjoy sensitivity and reproducibility in real-time PCR applications with Thermo Scientific™ ABsolute™ Probe-Based Detection qPCR Master Mixes.

  • High sensitivity & specificity - The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
  • Reproducibility - All mixes undergo extensive QC to ensure batch to batch consistency
  • Optimized buffer - For maximal performance in multiplex reactions
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
• 1 x 5 mL ABsolute QPCR ROX Mix (2X)

Store at -20°C
High
Taq DNA Polymerase
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
qPCR Mix
400 Reactions
2X
400 Reactions
5 mL
Primer-probe
qPCR
Standard
ABsolute
400 reactions
Gene Expression
2 X
DNA, dsDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.