Learn More
Thermo Scientific™ ABsolute QPCR Mix, SYBR Green, no ROX
Optimized for SYBR Green chemistry and contains all the components necessary to perform quantitative PCR, with the exception of template and primers.
£1916.00
Specifications
Detection Method | SYBR |
---|---|
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Reaction Speed | Standard |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11827692
|
Thermo Scientific™
AB1158B |
1,600 Reactions |
£1916.00
1600 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets.
ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, Fluorescein or no reference dye. A master mix optimized for capillary instruments is also available.
- Thermo Scientific ABsolute qPCR SYBR Green Mixes are optimized for SYBR Green chemistry and contain all the components necessary to perform quantitative PCR, with the exception of template and primers
- The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets
- ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, low ROX concentration, fluorescein or no reference dye
- A master mix optimized for capillary instruments is also available
Highlights
- High Sensitivity & Specificity – The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
- Optimised buffer containing SYBR Green I
- Reproducibility – All master mixes undergo extensive QC to ensure batch to batch consistency
- Store at -20°C
Specifications
SYBR | |
qPCR | |
Standard | |
ABsolute | |
Gene Expression | |
2X |
High | |
Taq DNA Polymerase | |
BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene | |
Real Time PCR SYBR Master Mix | |
qPCR Cyclers | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.