Learn More
Thermo Scientific™ ABsolute QPCR Mix, SYBR Green, ROX
Shop All Thermo Scientific ProductsDescription
The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets.
ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, Fluorescein or no reference dye. A master mix optimized for capillary instruments is also available.
- High Sensitivity & Specificity - The master mixes contain the patented hot start enzyme Thermo-Start DNA Polymerase
- Optimized buffer containing SYBR Green
- Reproducibility - All master mixes undergo extensive QC to ensure batch to batch consistency
- Broad dynamic range
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | Contains: • 1 x 5 mL ABsolute QPCR SYBR Green ROX Mix (2X) • 100 μL MgCl2 (1M) Sufficient for 400 x 25 μL reactions. Store at -20°C |
| Detection Method | SYBR |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | Taq DNA Polymerase |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System |
| Product Line | ABsolute |
| Product Type | Real Time PCR SYBR Master Mix |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.