missing translation for 'onlineSavingsMsg'
Learn More

Acarbose, Tocris Bioscience™

Product Code. 18084202
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Glucosidase α inhibitor (intestinal)

Inhibitor of intestinal α-glucosidase (IC50 = 11 nM). Antidiabetic; inhibits the hydrolysis of complex carbohydrates.

Chemical Identifiers

CAS 56180-94-0
Molecular Formula C25H43NO18
Molecular Weight (g/mol) 645.608
InChI Key XUFXOAAUWZOOIT-AKWDJIIVSA-N
Synonym acarbose
PubChem CID 131674886
IUPAC Name (3S,5S)-5-[(2R,3S,5S)-5-[(2R,3S,5S)-3,4-dihydroxy-6-methyl-5-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino]oxan-2-yl]oxy-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,4-triol
SMILES CC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)NC4C=C(C(C(C4O)O)O)CO

Specifications

Quantity 50 mg
Formula Weight 645.25
Percent Purity >98%
Chemical Name or Material Acarbose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.