Learn More
Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers
Record who opened the ultra low freezer door, when it was opened and how long it was open with access key options.
Brand: Thermo Scientific™ FRAC34567
This item is not returnable.
View return policy
Description
Analyze records of which staff opened the freezer door, what time is was opened and for what duration with the Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers.
Features
- Card access comes with 5 key cards
- Field installed - purchased separately
- Factory installed in the plant with purchase of unit
Specifications
Field installed | |
Revco™ UxF | |
Access Key Option; Card access control w/five key cards |
Product Content Correction
Product Title
Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers >
Spot an opportunity for improvement?Share a Content Correction