missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine™ System Accessories
Dovetail your SymLine™ System to suit your specific needs! Here, you will find a lot of useful ancillaries.
Brand: SymLine™ 106423
18.46 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- SymLine™ offers many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables to allow you to customize your SymLine™ system
- All accessories are electrically dissipative and approved for use in laboratories with explosion hazards
Specifications
Black | |
SymLine™ System Customization | |
HDPE-EC |
Blind Plug | |
32 mm dia. Pipe | |
Blind Plug |