missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine™ System Accessories
Dovetail your SymLine™ System to suit your specific needs! Here, you will find a lot of useful ancillaries.
Brand: SymLine™ 106611
163.46 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.17000kg
Description
- SymLine™ offers many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables to allow you to customize your SymLine™ system
- All accessories are electrically dissipative and approved for use in laboratories with explosion hazards
Specifications
GL25 (m) to GL25 (m) | |
10 mm | |
SymLine™ System Customization | |
HDPE-EC |
Black | |
Angled Cabinet Feed-through, Counter Nut, Two Silicone Washers | |
Cabinet with 28 mm Borehole and 120 mm Wall Thickness | |
Mounting Bracket |