missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine™ System Accessories
Dovetail your SymLine™ System to suit your specific needs! Here, you will find a lot of useful ancillaries.
Brand: SymLine™ 108042-S1
143.04 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- SymLine™ offers many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables to allow you to customize your SymLine™ system
- All accessories are electrically dissipative and approved for use in laboratories with explosion hazards
Specifications
S60/61 | |
Black | |
Canister with Integrated Welded Sensor Holder, Screwable Sleeve for Sensor | |
SCAT™ SafetySystem Funnels | |
Cabinet Feed-through |
10 L | |
185 x 290 x 265 mm | |
SymLine™ System Customization | |
HDPE-EC |