missing translation for 'onlineSavingsMsg'
Learn More

SymLine™ System Accessories

Dovetail your SymLine™ System to suit your specific needs! Here, you will find a lot of useful ancillaries.

Brand:  SymLine™ 108270

31.48 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.



Additional Details : Weight : 0.06000kg

 View more versions of this product

Product Code. 17307429

  • £39.95 / Each
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

  • SymLine™ offers many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables to allow you to customize your SymLine™ system
  • All accessories are electrically dissipative and approved for use in laboratories with explosion hazards
Specifications

Specifications

Yellow
Grounding Cable, Clamp, Clip
SCAT™ FlexTubes
3 m
SymLine™ System Customization
Grounding Cable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers