Learn More
SymLine™ System Accessories
Dovetail your SymLine™ System to suit your specific needs! Here, you will find a lot of useful ancillaries.
Brand:  SymLine™ 106608
85.83 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17337419
  • £108.00 / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • SymLine™ offers many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables to allow you to customize your SymLine™ system
  • All accessories are electrically dissipative and approved for use in laboratories with explosion hazards
Specifications
Specifications
GL25 (m) to GL25 (m)
10 mm
SymLine™ System Customization
HDPE-EC
Black
Cabinet Feed-through, Counter Nut, Two Silicone Washers
Cabinet with 28 mm Borehole and 120 mm Wall Thickness
Cabinet Feed-through