missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine™ System Accessories
Dovetail your SymLine™ System to suit your specific needs! Here, you will find a lot of useful ancillaries.
Brand: SymLine™ 108176
31.78 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.39000kg
Description
- SymLine™ offers many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables to allow you to customize your SymLine™ system
- All accessories are electrically dissipative and approved for use in laboratories with explosion hazards
Specifications
Yellow | |
Grounding Cable, Clamp, Clip | |
32 mm Dia. Pipe |
3 m | |
SymLine™ System Customization | |
Grounding Cable |