missing translation for 'onlineSavingsMsg'
Learn More

SymLine™ System Accessories

Dovetail your SymLine™ System to suit your specific needs! Here, you will find a lot of useful ancillaries.

Brand:  SymLine™ 108268

38.39 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 17883520

  • £47.90 / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • SymLine™ offers many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables to allow you to customize your SymLine™ system
  • All accessories are electrically dissipative and approved for use in laboratories with explosion hazards
Specifications

Specifications

Yellow
Grounding Cable, Clamp, Push Button
Solvent System
Grounding Cable
2 m
SymLine™ System Customization
6.6 ft.
Product Suggestions

Product Suggestions

Videos
SDS
Documentos

Documentos

Product Certifications
Ofertas especiales

Ofertas especiales