missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Accessories for Horizontal Gel Box, 10-20 cm

For use with horizontal gel box, 10cm.

Brand:  Axygen HGB10-16-1

 View more versions of this product

Product Code. 15553107

  • £43.25 / 1 unit
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Combs for 10-20 cm Axygen™ horizontal gel box
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Horizontal Electrophoresis System Accessories
16-well Comb, 1.0 mm thickness
1/Cs.
1 mm
Axygen Horizontal Gel Box, 10 cm
16
1/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers