missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Accessories for Horizontal Gel Box, 10-20 cm

For use with horizontal gel box, 10cm.

Brand:  Axygen HGB10-20MC-15

 View more versions of this product

Product Code. 15593107

  • £46.90 / 1 unit
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Combs for 10-20 cm Axygen™ horizontal gel box
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Horizontal Electrophoresis System Accessories
20-well Multi-Channel Compatible Comb, 1.5 mm thickness
1/Cs.
1.5 mm
Axygen Horizontal Gel Box, 10 cm
20
1/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers