Learn More
Thermo Scientific™ Accessories for Digital Heating Cooling Drybath
Optimize the performance of our digital heating cooling drybath and digital heating shaking bath by using these accessories.
£155.96 - £256.82
Product Code | Brand | No. of Places | Tube Size | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Places | Tube Size | Type | Price | Quantity & Availability | |||
15258884
|
Thermo Scientific™
88880129 |
48 | 0.5mL | Block |
£155.96
Each |
In Stock
Log in to see stock. |
|||
15268884
|
Thermo Scientific™
88880130 |
30 | 1.5mL | Block |
£155.96
Each |
In Stock
Log in to see stock. |
|||
15278884
|
Thermo Scientific™
88880131 |
15 | 15mL | Block |
£155.96
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||
15288884
|
Thermo Scientific™
88880132 |
6 | 50mL small | Block |
£189.86
Each |
In Stock
Log in to see stock. |
|||
15298884
|
Thermo Scientific™
88880133 |
6 | 50mL large | Block |
£256.82
Each |
In Stock
Log in to see stock. |
|||
15208894
|
Thermo Scientific™
88880134 |
35 | 10mm | Block |
£155.96
Each |
In Stock
Log in to see stock. |
|||
15218894
|
Thermo Scientific™
88880135 |
24 | 12mm | Block |
£155.96
Each |
In Stock
Log in to see stock. |
|||
15228894
|
Thermo Scientific™
88880136 |
24 | 13mm | Block |
£155.96
Each |
In Stock
Log in to see stock. |
|||
15248894
|
Thermo Scientific™
88880137 |
20 | 15mm | Block |
£155.96
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||
15258894
|
Thermo Scientific™
88880138 |
16 | 16mm | Block |
£155.96
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||
15268894
|
Thermo Scientific™
88880139 |
12 | 18mm | Block |
£155.96
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||
15278894
|
Thermo Scientific™
88880140 |
12 | 20mm | Block |
£155.96
Each |
In Stock
Log in to see stock. |
|||
Description
Use Thermo Scientific™ Accessories for Digital Heating Cooling Bath to optimize flexibility and performance.
- Warranty: 2 years
Specifications
LED | |
Block |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.